Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EWSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039267.2 | NP_001034356.2 | 13 - 14 | 1463 | 134 | |
XM_015294853.2 | XP_015150339.1 | 14 - 15 | 1469 | 134 | ||
XM_015294854.2 | XP_015150340.1 | 14 - 15 | 1466 | 134 | ||
XM_015294855.2 | XP_015150341.1 | 13 - 14 | 1466 | 134 | ||
GenBank mRNA | AJ719366.1 | - | 13 - 14 | 1463 | 134 | |
CD740517.1 | - | 2 - 3 | 268 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |