Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EWSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039267.2 | NP_001034356.2 | 9 - 10 | 1049 | 87 | |
XM_015294853.2 | XP_015150339.1 | 10 - 11 | 1055 | 87 | ||
XM_015294854.2 | XP_015150340.1 | 10 - 11 | 1052 | 87 | ||
XM_015294855.2 | XP_015150341.1 | 9 - 10 | 1052 | 87 | ||
GenBank mRNA | AJ719366.1 | - | 9 - 10 | 1049 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |