Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039318.1 | NP_001034407.1 | 7 - 8 | 2065 | 59 | |
XM_015296936.2 | XP_015152422.1 | 12 - 13 | 3945 | 59 | ||
XM_015296939.2 | XP_015152425.1 | 11 - 12 | 3834 | 59 | ||
XM_025142546.1 | XP_024998314.1 | 12 - 13 | 3948 | 59 | ||
XM_025142547.1 | XP_024998315.1 | 11 - 12 | 3837 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |