Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025149682.1 | XP_025005450.1 | 3 - 4 | 703 | 142 | |
GenBank mRNA | AY633575.2 | - | 2 - 3 | 595 | 142 | |
KM486597.1 | - | 2 - 3 | 451 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |