Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032390.2 | NP_001027562.2 | 2 - 3 | 158 | 104 | |
GenBank mRNA | BX931608.2 | - | 2 - 3 | 142 | 104 | |
BX933641.1 | - | 2 - 3 | 133 | 104 | ||
BX933648.1 | - | 2 - 3 | 140 | 104 | ||
BX933649.2 | - | 2 - 3 | 140 | 104 | ||
BX933653.2 | - | 2 - 3 | 155 | 104 | ||
BX933662.1 | - | 2 - 3 | 123 | 104 | ||
BX933671.1 | - | 2 - 3 | 133 | 104 | ||
BX933680.2 | - | 2 - 3 | 132 | 104 | ||
BX933686.1 | - | 2 - 3 | 158 | 104 | ||
BX933689.1 | - | 2 - 3 | 152 | 104 | ||
BX935221.1 | - | 2 - 3 | 128 | 104 | ||
BX935254.1 | - | 2 - 3 | 168 | 104 | ||
BX935312.1 | - | 2 - 3 | 130 | 104 | ||
BX935323.2 | - | 2 - 3 | 140 | 104 | ||
BX935324.2 | - | 2 - 3 | 132 | 104 | ||
BX935330.2 | - | 2 - 3 | 120 | 104 | ||
BX936261.2 | - | 2 - 3 | 159 | 104 | ||
CR338993.2 | - | 2 - 3 | 140 | 104 | ||
CR338996.1 | - | 2 - 3 | 142 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |