Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL10RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039597.1 | NP_001034686.1 | 3 - 4 | 387 | 71 | |
GenBank mRNA | AM049243.1 | - | 3 - 4 | 387 | 71 | |
AM049244.1 | - | 3 - 4 | 387 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |