Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039601.1 | NP_001034690.1 | 7 - 8 | 648 | 76 | |
XM_025152179.1 | XP_025007947.1 | 8 - 9 | 702 | 76 | ||
XM_025152180.1 | XP_025007948.1 | 5 - 6 | 515 | 76 | ||
GenBank mRNA | DQ150103.1 | - | 7 - 8 | 648 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |