Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044645.1 | NP_001038110.1 | 2 - 3 | 283 | 132 | |
XM_025144266.1 | XP_025000034.1 | 2 - 3 | 132 | |||
GenBank mRNA | AY531258.1 | - | 2 - 3 | 263 | 132 | |
AY531259.1 | - | 2 - 3 | 263 | 132 | ||
AY531260.1 | - | 2 - 3 | 263 | 132 | ||
AY531261.1 | - | 2 - 3 | 263 | 132 | ||
CR339010.1 | - | 2 - 3 | 256 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Gallus gallus Probe spans exons |