Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044671.1 | NP_001038136.1 | 1 - 2 | 1650 | 67 | |
XM_015292514.2 | XP_015148000.1 | 1 - 2 | 1523 | 67 | ||
GenBank mRNA | D28598.1 | - | 1 - 2 | 1650 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |