Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045829.1 | NP_001039294.1 | 3 - 4 | 341 | 74 | |
XM_025156060.1 | XP_025011828.1 | 3 - 4 | 329 | 74 | ||
GenBank mRNA | BX931905.1 | - | 3 - 4 | 384 | 74 | |
BX934989.1 | - | 3 - 4 | 383 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |