Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZPAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045837.1 | NP_001039302.1 | 8 - 9 | 1039 | 149 | |
XM_015284957.1 | XP_015140443.1 | 10 - 11 | 1208 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |