Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048007.1 | NP_001041472.1 | 5 - 6 | 475 | 60 | |
XM_015289171.2 | XP_015144657.1 | 5 - 6 | 492 | 60 | ||
GenBank mRNA | BX929892.1 | - | 5 - 6 | 475 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |