Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079575.1 | NP_001073043.1 | 5 - 6 | 733 | 76 | |
GenBank mRNA | AB284187.1 | - | 5 - 6 | 733 | 76 | |
CR390660.1 | - | 4 - 5 | 581 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |