Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080104.1 | NP_001073573.1 | 4 - 5 | 450 | 87 | |
XM_025143973.1 | XP_024999741.1 | 4 - 5 | 475 | 87 | ||
GenBank mRNA | BX934948.2 | - | 3 - 4 | 403 | 87 | |
BX934959.1 | - | 4 - 5 | 437 | 87 | ||
EF010531.1 | - | 4 - 5 | 450 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons |