Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNUPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080878.1 | NP_001074347.1 | 6 - 7 | 712 | 149 | |
GenBank mRNA | AJ720941.1 | - | 6 - 7 | 712 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |