Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABEPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080891.1 | NP_001074360.1 | 6 - 7 | 899 | 110 | |
GenBank mRNA | AJ720597.1 | - | 6 - 7 | 899 | 110 | |
CR406324.1 | - | 4 - 5 | 629 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |