Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081696.1 | NP_001075165.1 | 3 - 4 | 594 | 83 | |
GenBank mRNA | AY069983.1 | - | 2 - 3 | 325 | 83 | |
AY753288.1 | - | 2 - 3 | 435 | 83 | ||
EF068233.1 | - | 3 - 4 | 594 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |