Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BG-like antigen 1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099355.1 | NP_001092825.1 | 9 - 10 | 672 | 151 | |
XM_015294968.2 | XP_015150454.1 | 9 - 10 | 1177 | 238 | ||
XM_015294970.2 | XP_015150456.1 | 9 - 10 | 1173 | 238 | ||
XM_015294971.2 | XP_015150457.1 | 9 - 10 | 1177 | 235 | ||
XM_015294972.2 | XP_015150458.1 | 9 - 10 | 1178 | 235 | ||
XM_015294973.2 | XP_015150459.1 | 8 - 9 | 1156 | 238 | ||
XM_015294974.2 | XP_015150460.1 | 8 - 9 | 1152 | 238 | ||
XM_015294977.2 | XP_015150463.1 | 9 - 10 | 1174 | 151 | ||
XM_015294979.2 | XP_015150465.1 | 9 - 10 | 1175 | 148 | ||
XM_015294981.2 | XP_015150467.1 | 9 - 10 | 1171 | 148 | ||
GenBank mRNA | KC955132.1 | - | 9 - 10 | 151 | ||
KC955133.1 | - | 10 - 11 | 151 | |||
KC955134.1 | - | 10 - 11 | 151 | |||
KC955135.1 | - | 10 - 11 | 151 | |||
KC955136.1 | - | 9 - 10 | 151 | |||
KX686501.1 | - | 9 - 10 | 151 | |||
KX686503.1 | - | 9 - 10 | 907 | 238 | ||
KX686505.1 | - | 9 - 10 | 902 | 238 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |