Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSMCE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105064.1 | NP_001098534.1 | 437 | 68 | ||
GenBank mRNA | BX930943.2 | - | 301 | 68 | ||
BX932334.2 | - | 437 | 68 | |||
BX934453.2 | - | 301 | 68 | |||
CR354280.1 | - | 342 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus |