Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105610.1 | NP_001099080.1 | 1 - 2 | 70 | 90 | |
GenBank mRNA | BX930529.2 | - | 2 - 3 | 193 | 90 | |
BX933927.2 | - | 2 - 3 | 251 | 90 | ||
EU069394.1 | - | 1 - 2 | 70 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |