Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105675.1 | NP_001099145.1 | 2 - 3 | 286 | 101 | |
GenBank mRNA | AJ720942.1 | - | 2 - 3 | 286 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |