Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205148.4 | NP_990479.1 | 1 - 2 | 1206 | 80 | |
XM_015284820.2 | XP_015140306.1 | 2 - 3 | 685 | 80 | ||
GenBank mRNA | S83278.2 | - | 1 - 2 | 567 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |