Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205520.4 | NP_990851.1 | 4 - 5 | 697 | 95 | |
GenBank mRNA | J02787.1 | - | 4 - 5 | 697 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |