Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113289.1 | NP_001106760.1 | 2 - 3 | 242 | 103 | |
XM_015287039.2 | XP_015142525.1 | 3 - 4 | 333 | 103 | ||
XM_015287040.2 | XP_015142526.1 | 3 - 4 | 376 | 103 | ||
XM_015287041.2 | XP_015142527.1 | 3 - 4 | 400 | 103 | ||
GenBank mRNA | AF419846.1 | - | 3 - 4 | 351 | 103 | |
M61754.1 | - | 2 - 3 | 269 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |