Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113293.1 | NP_001106764.1 | 2 - 3 | 375 | 82 | |
GenBank mRNA | EU165362.1 | - | 2 - 3 | 375 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |