Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113571.1 | NP_001107043.1 | 4 - 5 | 569 | 129 | |
GenBank mRNA | BI391443.1 | - | 4 - 5 | 539 | 129 | |
CR352757.1 | - | 4 - 5 | 572 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |