Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204112.1 | NP_989443.1 | 4 - 5 | 829 | 146 | |
XM_025152725.1 | XP_025008493.1 | 4 - 5 | 767 | 146 | ||
GenBank mRNA | AF532926.1 | - | 4 - 5 | 829 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |