Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TWSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204198.1 | NP_989529.1 | 3 - 4 | 285 | 73 | |
XM_025146996.1 | XP_025002764.1 | 3 - 4 | 306 | 73 | ||
GenBank mRNA | AF255731.1 | - | 3 - 4 | 285 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |