Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204199.1 | NP_989530.1 | 7 - 8 | 736 | 83 | |
XM_015280581.2 | XP_015136067.1 | 7 - 8 | 758 | 83 | ||
XM_015280582.2 | XP_015136068.1 | 7 - 8 | 758 | 83 | ||
GenBank mRNA | AF322650.1 | - | 7 - 8 | 736 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |