Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPD52L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204215.1 | NP_989546.1 | 5 - 6 | 402 | 77 | |
XM_015284431.2 | XP_015139917.1 | 6 - 7 | 535 | 77 | ||
GenBank mRNA | AY004870.1 | - | 5 - 6 | 402 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |