Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204288.1 | NP_989619.1 | 7 - 8 | 1130 | 67 | |
XM_015292934.2 | XP_015148420.1 | 7 - 8 | 1113 | 67 | ||
XM_015292935.1 | XP_015148421.1 | 7 - 8 | 1113 | 67 | ||
XM_025154402.1 | XP_025010170.1 | 7 - 8 | 1113 | 67 | ||
XM_025154403.1 | XP_025010171.1 | 7 - 8 | 1166 | 67 | ||
GenBank mRNA | AF439559.1 | - | 7 - 8 | 1130 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |