Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204379.2 | NP_989710.2 | 3 - 4 | 360 | 146 | |
XM_015291626.2 | XP_015147112.1 | 4 - 5 | 731 | 146 | ||
XM_015291627.2 | XP_015147113.1 | 4 - 5 | 727 | 146 | ||
XM_015291628.2 | XP_015147114.1 | 3 - 4 | 333 | 146 | ||
GenBank mRNA | AB114678.1 | - | 3 - 4 | 339 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |