Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204384.1 | NP_989715.1 | 4 - 5 | 458 | 71 | |
XM_015273097.2 | XP_015128583.1 | 4 - 5 | 1162 | 71 | ||
GenBank mRNA | AB004542.1 | - | 4 - 5 | 508 | 71 | |
AY298725.1 | - | 4 - 5 | 458 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |