Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204391.1 | NP_989722.1 | 5 - 6 | 495 | 75 | |
XM_015283077.2 | XP_015138563.1 | 6 - 7 | 697 | 75 | ||
XM_015283079.2 | XP_015138565.1 | 6 - 7 | 682 | 75 | ||
XM_025147006.1 | XP_025002774.1 | 5 - 6 | 660 | 75 | ||
GenBank mRNA | AJ721107.1 | - | 5 - 6 | 679 | 75 | |
AY278230.1 | - | 5 - 6 | 495 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |