Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GZMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204457.1 | NP_989788.1 | 4 - 5 | 613 | 97 | |
GenBank mRNA | AJ544060.1 | - | 4 - 5 | 613 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |