Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204461.1 | NP_989792.1 | 4 - 5 | 492 | 138 | |
GenBank mRNA | AF372468.1 | - | 4 - 5 | 492 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |