Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS2ST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204481.1 | NP_989812.1 | 2 - 3 | 530 | 144 | |
GenBank mRNA | AB093516.1 | - | 2 - 3 | 530 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |