Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204485.2 | NP_989816.2 | 18 - 19 | 2581 | 64 | |
XM_015277227.2 | XP_015132713.1 | 18 - 19 | 2493 | 64 | ||
GenBank mRNA | AB109635.1 | - | 17 - 18 | 2426 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |