Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204496.1 | NP_989827.1 | 1 - 2 | 266 | 71 | |
XM_015293723.2 | XP_015149209.2 | 2 - 3 | 255 | 71 | ||
XM_015293725.2 | XP_015149211.2 | 1 - 2 | 263 | 71 | ||
XM_015293730.2 | XP_015149216.2 | 1 - 2 | 266 | 71 | ||
XM_025154783.1 | XP_025010551.1 | 1 - 2 | 266 | 71 | ||
XM_025154784.1 | XP_025010552.1 | 2 - 3 | 235 | 71 | ||
XM_025154785.1 | XP_025010553.1 | 1 - 2 | 266 | 71 | ||
XM_025154787.1 | XP_025010555.1 | 1 - 2 | 269 | 71 | ||
XM_025154788.1 | XP_025010556.1 | 1 - 2 | 266 | 71 | ||
XM_025154789.1 | XP_025010557.1 | 1 - 2 | 296 | 71 | ||
XM_025154790.1 | XP_025010558.1 | 2 - 3 | 192 | 71 | ||
XM_025154795.1 | XP_025010563.1 | 1 - 2 | 266 | 71 | ||
XM_025154796.1 | XP_025010564.1 | 2 - 3 | 184 | 71 | ||
XM_025154797.1 | XP_025010565.1 | 1 - 2 | 263 | 71 | ||
GenBank mRNA | AY278548.1 | - | 1 - 2 | 266 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |