Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204500.1 | NP_989831.1 | 3 - 4 | 622 | 89 | |
XM_015297942.2 | XP_015153428.1 | 3 - 4 | 2893 | 89 | ||
XM_025143169.1 | XP_024998937.1 | 3 - 4 | 543 | 89 | ||
GenBank mRNA | AF257773.1 | - | 3 - 4 | 622 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |