Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204513.1 | NP_989844.1 | 4 - 5 | 561 | 78 | |
XM_015284597.2 | XP_015140083.1 | 4 - 5 | 660 | 78 | ||
GenBank mRNA | AJ535486.1 | - | 4 - 5 | 561 | 78 | |
BX929414.2 | - | 4 - 5 | 635 | 78 | ||
BX930442.2 | - | 4 - 5 | 570 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |