Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204514.2 | NP_989845.2 | 6 - 7 | 747 | 62 | |
GenBank mRNA | AJ535485.1 | - | 740 | 62 | ||
BX931363.1 | - | 5 - 6 | 597 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |