Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205176.1 | NP_990507.1 | 1 - 2 | 220 | 78 | |
GenBank mRNA | BX930014.2 | - | 1 - 2 | 235 | 78 | |
S53164.1 | - | 1 - 2 | 220 | 78 | ||
U26661.1 | - | 1 - 2 | 400 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |