Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205205.2 | NP_990536.2 | 2 - 3 | 494 | 91 | |
XM_015281306.2 | XP_015136792.1 | 3 - 4 | 497 | 91 | ||
XM_015281307.2 | XP_015136793.1 | 3 - 4 | 497 | 91 | ||
GenBank mRNA | CR382426.1 | - | 2 - 3 | 494 | 91 | |
X80258.1 | - | 424 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |