Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205267.1 | NP_990598.1 | 7 - 8 | 634 | 189 | |
GenBank mRNA | BX935061.2 | - | 7 - 8 | 617 | 189 | |
X17200.1 | - | 634 | 189 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |