Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005616.1 | NP_001005616.1 | 2 - 3 | 1904 | 79 | |
XM_015283620.2 | XP_015139106.1 | 3 - 4 | 2288 | 79 | ||
XM_015283621.2 | XP_015139107.1 | 3 - 4 | 2415 | 79 | ||
XM_015283622.2 | XP_015139108.1 | 3 - 4 | 2284 | 79 | ||
GenBank mRNA | U46563.1 | - | 2 - 3 | 1904 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |