Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006140.2 | NP_001006140.2 | 7 - 8 | 765 | 90 | |
GenBank mRNA | AJ720805.1 | - | 7 - 8 | 766 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons |