Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006142.1 | NP_001006142.1 | 5 - 6 | 563 | 101 | |
XM_025154495.1 | XP_025010263.1 | 8 - 9 | 6316 | 101 | ||
GenBank mRNA | AJ719288.1 | - | 5 - 6 | 563 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |