Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006172.2 | NP_001006172.1 | 2 - 3 | 482 | 150 | |
XM_015294230.2 | XP_015149716.1 | 2 - 3 | 476 | 150 | ||
GenBank mRNA | AJ719408.1 | - | 2 - 3 | 484 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |