Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006177.1 | NP_001006177.1 | 1 - 2 | 93 | 82 | |
XM_025148593.1 | XP_025004361.1 | 1 - 2 | 146 | 82 | ||
GenBank mRNA | AJ720833.1 | - | 1 - 2 | 93 | 82 | |
CR389678.1 | - | 1 - 2 | 92 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |